Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Acryloylamido-Ethoxyethanol Solution, ≥98.0%, Honeywell™ Fluka™

Click to view available options
Quantity:
1mL
5mL
Packaging:
Glass Bottle
89911-50-2
C7H13NO3
159.185
MFCD11975206
BYJPRUDFDZPCBH-UHFFFAOYSA-N
n-acryloylamido-ethoxyethanol solution,n-2-2-hydroxyethoxy ethyl prop-2-enamide,n-acryloylamido-ethoxyethanol,2-2-acryloylamino ethoxy ethanol,n-2-hydroxyethoxy ethyl-acrylamide,n-2-2-hydroxyethoxy ethyl acrylamide
19364541
N-[2-(2-hydroxyethoxy)ethyl]prop-2-enamide
C=CC(=O)NCCOCCO
89911-50-2
C7H13NO3
159.185
MFCD11975206
BYJPRUDFDZPCBH-UHFFFAOYSA-N
n-acryloylamido-ethoxyethanol solution,n-2-2-hydroxyethoxy ethyl prop-2-enamide,n-acryloylamido-ethoxyethanol,2-2-acryloylamino ethoxy ethanol,n-2-hydroxyethoxy ethyl-acrylamide,n-2-2-hydroxyethoxy ethyl acrylamide
19364541
N-[2-(2-hydroxyethoxy)ethyl]prop-2-enamide
C=CC(=O)NCCOCCO
Concentration or Composition (by Analyte or Components) approx.50% in water
Quantity 1mL
Color Red
Packaging Glass Bottle
Refractive Index n20/D 1.416
Density 1.075g/mL at 20°C
Appearance Viscous liquid
UN Number NONH for all modes of transport
Formula Weight 159.18g/mol
Percent Purity ≥98.0% (HPLC)
Chemical Name or Material N-Acryloylamido-Ethoxyethanol Solution
Show More Show Less