Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Allylaniline, 95%

Manufacturer:  Alfa Aesar™ H5582106

Quantity 5g

 View more versions of this product

Catalog No. AAH5582106


  Check Availability
Add to cart

Specifications

Description

Applications
It is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Keep away from oxidizing agents and strong acids.
Description & Specifications

Chemical Identifiers

589-09-3
133.19
N-Allylaniline
C6H5NHCH2CH=CH2
MFCD00008638

Specifications

218° to 220°C
5g
0.982g/mL
Liquid
133.19
N-Allylaniline
Colorless to brown
589-09-3
1.5630
C6H5NHCH2CH=CH2
MFCD00008638
95%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H301-H311-H227-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Combustible liquid.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P301+P310-P305+P351+P338-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit