Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Allylbenzothiazolium Bromide 98.0+%, TCI America™

Click to view available options
Quantity:
25g
16407-55-9
C10H10BrNS
256.161
MFCD00191299
MEYXQRPXVJWXBZ-UHFFFAOYSA-M
177743
3-prop-2-enyl-1,3-benzothiazol-3-ium;bromide
C=CC[N+]1=CSC2=CC=CC=C21.[Br-]
16407-55-9
C10H10BrNS
256.161
MFCD00191299
MEYXQRPXVJWXBZ-UHFFFAOYSA-M
177743
3-prop-2-enyl-1,3-benzothiazol-3-ium;bromide
C=CC[N+]1=CSC2=CC=CC=C21.[Br-]
Melting Point 150°C
Quantity 25g
Color Undesignated
Formula Weight 256.16
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (HPLC,T)
Chemical Name or Material N-Allylbenzothiazolium Bromide

EINECSNumber : (5)-0233

TSCA : No