Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-alpha-Benzyloxycarbonyl-N-epsilon-Boc-L-lysine, 99%

Manufacturer:  Alfa Aesar™ B2201003

Quantity 1g

 View more versions of this product

Catalog No. AAB2201003


  Check Availability
Add to cart

Specifications

Description

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Soluble in DMF (0.5 mmol/ml).

Notes
Store at -20°C. Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
Description & Specifications

Chemical Identifiers

2389-60-8
380.44
N-Benzyloxycarbonyl-N-epsilon-tert-butoxycarbonyl-L-lysine; N-Cbz-N-epsilon-Boc-L-Lysine
C19H28N2O6
MFCD00062271
N-alpha-Benzyloxycarbonyl-N-epsilon-Boc-L-lysine

Specifications

2389-60-8
380.44
N-Benzyloxycarbonyl-N-epsilon-tert-butoxycarbonyl-L-lysine; N-Cbz-N-epsilon-Boc-L-Lysine
1g
99%
C19H28N2O6
MFCD00062271
64° to 67°C
N-alpha-Benzyloxycarbonyl-N-epsilon-Boc-L-lysine
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit