Alfa Aesar™ N-alpha-Benzyloxycarbonyl-N-epsilon-Boc-L-lysine, 99%

Manufacturer: Alfa Aesar™ B2201006

Quantity 5g

 View more versions of this product

Catalog No. AAB2201006

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2389-60-8
    Molecular Formula C19H28N2O6
    Formula Weight 380.44
    MDL Number MFCD00062271
    Synonym N-Benzyloxycarbonyl-N-epsilon-tert-butoxycarbonyl-L-lysine; N-Cbz-N-epsilon-Boc-L-Lysine
    Chemical Name or Material N-alpha-Benzyloxycarbonyl-N-epsilon-Boc-L-lysine

    Specifications

    Formula Weight 380.44
    Molecular Formula C19H28N2O6
    MDL Number MFCD00062271
    CAS 2389-60-8
    Synonym N-Benzyloxycarbonyl-N-epsilon-tert-butoxycarbonyl-L-lysine; N-Cbz-N-epsilon-Boc-L-Lysine
    Melting Point 64° to 67°C
    Quantity 5g
    Chemical Name or Material N-alpha-Benzyloxycarbonyl-N-epsilon-Boc-L-lysine
    Assay 99%

    Description

    Applications
    It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

    Solubility
    Soluble in DMF (0.5 mmol/ml).

    Notes
    Store at -20°C. Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit