Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N(alpha)-Boc-D-tryptophanol, 98%

Manufacturer:  Alfa Aesar™ H2776306

Quantity 5g

 View more versions of this product

Catalog No. AAH2776306


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

158932-00-4
290.36
Boc-D-Trp-ol
C16H22N2O3
MFCD00270221
N(alpha)-Boc-D-tryptophanol

Specifications

158932-00-4
5g
C16H22N2O3
MFCD00270221
N(alpha)-Boc-D-tryptophanol
119° to 121°C
+53° (c=2 in Acetic Acid)
290.36
Boc-D-Trp-ol
98%
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit