Alfa Aesar™ N(alpha)-Boc-L-arginine, 98%

Manufacturer: Alfa Aesar™ H5337906

Quantity 5g

 View more versions of this product

Catalog No. AAH5337906

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13726-76-6
    Molecular Formula C11H22N4O4
    Formula Weight 274.32
    MDL Number MFCD00042632
    Beilstein 2419628
    Synonym Boc-Arg-OH; Nalpha-tert-Butoxycarbonyl-L-arginine
    Chemical Name or Material N(alpha)-Boc-L-arginine

    Specifications

    Formula Weight 274.32
    Molecular Formula C11H22N4O4
    MDL Number MFCD00042632
    CAS 13726-76-6
    Beilstein 2419628
    Synonym Boc-Arg-OH; Nalpha-tert-Butoxycarbonyl-L-arginine
    Flash Point 113°C (235°F)
    Optical Rotation −6.5° (c=1 in Acetic Acid)
    Quantity 5g
    Chemical Name or Material N(alpha)-Boc-L-arginine
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit