Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N(alpha)-Boc-L-histidine, 98+%

Click to view available options
Quantity:
1g
5g
17791-52-5
C11H17N3O4
255.274
MFCD00065576
AYMLQYFMYHISQO-QMMMGPOBSA-N
boc-his-oh,n-boc-l-histidine,boc-l-histidine,tert-butyloxycarbonyl-l-histidine,nalpha-tert-butoxycarbonyl-l-histidine,nalpha-boc-l-histidine,boc-l-his-oh,n-tert-butoxycarbonyl-l-histidine,l-histidine, n-1,1-dimethylethoxy carbonyl
87308
(2S)-3-(1H-imidazol-5-yl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
CC(C)(C)OC(=O)NC(CC1=CN=CN1)C(=O)O
17791-52-5
C11H17N3O4
255.274
MFCD00065576
AYMLQYFMYHISQO-QMMMGPOBSA-N
boc-his-oh,n-boc-l-histidine,boc-l-histidine,tert-butyloxycarbonyl-l-histidine,nalpha-tert-butoxycarbonyl-l-histidine,nalpha-boc-l-histidine,boc-l-his-oh,n-tert-butoxycarbonyl-l-histidine,l-histidine, n-1,1-dimethylethoxy carbonyl
87308
(2S)-3-(1H-imidazol-5-yl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
CC(C)(C)OC(=O)NC(CC1=CN=CN1)C(=O)O
Beilstein 755289
Formula Weight 255.27
Percent Purity ≥98%
Melting Point ~198°C (decomposition)
Quantity 5g
Optical Rotation +26° (c=1 in Methanol)
Chemical Name or Material N(alpha)-Boc-L-histidine

EINECSNumber : 241-768-9

TSCA : No

Recommended Storage : Ambient temperatures