Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-α-BOC-N(im)-tosyl-L-histidine, 98%, Acros Organics™

Click to view available options
Quantity:
5g
25g
35899-43-5
C18H23N3O6S
409.46
DCLJSEPKYJSEHW-HNNXBMFYSA-N
boc-his tos-oh,boc-l-his tos-oh,boc-his tos,boc-l-histidine tosyl,s-2-tert-butoxycarbonyl amino-3-1-tosyl-1h-imidazol-4-yl propanoic acid,2s-2-tert-butoxycarbonyl amino-3-1-4-methylbenzenesulfonyl imidazol-4-yl propanoic acid,boc-his-tos,ambotzbaa1153,pubchem7363,boc-his-tos-oh
6455010
(2S)-3-[1-(4-methylphenyl)sulfonylimidazol-4-yl]-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
CC1=CC=C(C=C1)S(=O)(=O)N2C=C(N=C2)CC(C(=O)O)NC(=O)OC(C)(C)C
35899-43-5
C18H23N3O6S
409.46
DCLJSEPKYJSEHW-HNNXBMFYSA-N
boc-his tos-oh,boc-l-his tos-oh,boc-his tos,boc-l-histidine tosyl,s-2-tert-butoxycarbonyl amino-3-1-tosyl-1h-imidazol-4-yl propanoic acid,2s-2-tert-butoxycarbonyl amino-3-1-4-methylbenzenesulfonyl imidazol-4-yl propanoic acid,boc-his-tos,ambotzbaa1153,pubchem7363,boc-his-tos-oh
6455010
(2S)-3-[1-(4-methylphenyl)sulfonylimidazol-4-yl]-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
CC1=CC=C(C=C1)S(=O)(=O)N2C=C(N=C2)CC(C(=O)O)NC(=O)OC(C)(C)C
Quantity 25g
Melting Point 117.0°C to 118.0°C
Specific Rotation + 17.30
Assay Percent Range 97.5% min. (HPLC)
Formula Weight 409.46
Percent Purity 98%
Chemical Name or Material N-α-BOC-N(im)-tosyl-L-histidine