Alfa Aesar™ N(alpha)-Fmoc-D-tryptophan, 98%

Manufacturer: Alfa Aesar™ B2202214

Quantity 25g

 View more versions of this product

Catalog No. AAB2202214

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 86123-11-7
    Molecular Formula C26H22N2O4
    Formula Weight 426.48
    MDL Number MFCD00062954
    Synonym N-(9-Fluorenylmethoxycarbonyl)-D-tryptophan; Fmoc-D-Trp-OH
    Chemical Name or Material N(alpha)-Fmoc-D-tryptophan

    Specifications

    Formula Weight 426.48
    Molecular Formula C26H22N2O4
    MDL Number MFCD00062954
    CAS 86123-11-7
    Melting Point 182° to 185°C
    Quantity 25g
    Recommended Storage 2° to 8°C
    Synonym N-(9-Fluorenylmethoxycarbonyl)-D-tryptophan; Fmoc-D-Trp-OH
    Chemical Name or Material N(alpha)-Fmoc-D-tryptophan
    Assay 98%

    2° to 8°C

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit