Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N(alpha)-Fmoc-D-tryptophan, 98%

Click to view available options
Quantity:
1g
5g
25g
86123-11-7
C26H22N2O4
426.472
MFCD00062954
MGHMWKZOLAAOTD-XMMPIXPASA-N
fmoc-d-trp-oh,fmoc-d-tryptophan,n-9-fluorenylmethoxycarbonyl-d-tryptophan,n-alpha-fmoc-d-tryptophan,fmoc-d-trp,n-a-fmoc-d-tryptophan,nalpha-9h-fluoren-9-ylmethoxy carbonyl-d-tryptophan,2r-2-9h-fluoren-9-ylmethoxycarbonylamino-3-1h-indol-3-yl propanoic acid,r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-1h-indol-3-yl propanoic acid,d-tryptophan, n-9h-fluoren-9-ylmethoxy carbonyl
978344
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(1H-indol-3-yl)propanoic acid
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CNC5=CC=CC=C54)C(=O)O
86123-11-7
C26H22N2O4
426.472
MFCD00062954
MGHMWKZOLAAOTD-XMMPIXPASA-N
fmoc-d-trp-oh,fmoc-d-tryptophan,n-9-fluorenylmethoxycarbonyl-d-tryptophan,n-alpha-fmoc-d-tryptophan,fmoc-d-trp,n-a-fmoc-d-tryptophan,nalpha-9h-fluoren-9-ylmethoxy carbonyl-d-tryptophan,2r-2-9h-fluoren-9-ylmethoxycarbonylamino-3-1h-indol-3-yl propanoic acid,r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-1h-indol-3-yl propanoic acid,d-tryptophan, n-9h-fluoren-9-ylmethoxy carbonyl
978344
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(1H-indol-3-yl)propanoic acid
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CNC5=CC=CC=C54)C(=O)O
Formula Weight 426.48
Percent Purity 98%
Melting Point 182°C to 185°C
Quantity 1g
Chemical Name or Material N(alpha)-Fmoc-D-tryptophan

Recommended Storage : 2° to 8°C