Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Amyl Alcohol (Certified ACS), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.

Packaging Glass Bottle
Quantity 4L

 View more versions of this product

Catalog No. A394-4



  Check Availability
Add to cart

Custom packing/Bulk ordering

You can now order this item in custom quantities. Learn More

Specifications Description & Specifications

Chemical Identifiers

71-41-0
C5H12O
88.15
1-Pentanol
≥98.0%
≥98.0 %
CH3(CH2)4OH
MFCD00002977
n-Amyl Alcohol
Certified ACS Grade

Specifications

0.075mEq/gM max.
0.1% max.
0.003% max.
Liquid
≥98.0 %
Colorless
137°C
−78°C
MFCD00002977
1105
1-Pentanol
≥98.0%
71-41-0
Glass Bottle
0.5% max.
0.811g/cm³
C5H12O
CH3(CH2)4OH
88.15
7
4L
48°C
n-Amyl Alcohol
Certified ACS Grade

Safety and Handling

SDS & Safety Documents

DOTInformation : DOT Class 3, : Flammable Liquid

Alternative Products
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit