Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Arachidonoyldopamine, 98%

Click to view available options
Quantity:
5mg
25mg
199875-69-9
C28H41NO3
439.64
MFCD03412031
MVVPIAAVGAWJNQ-DOFZRALJSA-N
nada,arachidonoyl dopamine,n-arachidonoyl dopamine,arachidonyl dopamine,n-arachidonyldopamine,n-arachidonoyldopamine,n-arachidonyl dopamine,n-5z,8z,11z,14z-eicosatetraenoyl-dopamine,5z,8z,11z,14z-n-2-3,4-dihydroxyphenyl ethyl icosa-5,8,11,14-tetraenamide,n-2-3,4-dihydroxyphenyl ethyl-5z,8z,11z,14z-eicosatetraenamide
5282105
CHEBI:31231
(5Z,8Z,11Z,14Z)-N-[2-(3,4-dihydroxyphenyl)ethyl]icosa-5,8,11,14-tetraenamide
CCCCCC=CCC=CCC=CCC=CCCCC(=O)NCCC1=CC(=C(C=C1)O)O
199875-69-9
C28H41NO3
439.64
MFCD03412031
MVVPIAAVGAWJNQ-DOFZRALJSA-N
nada,arachidonoyl dopamine,n-arachidonoyl dopamine,arachidonyl dopamine,n-arachidonyldopamine,n-arachidonoyldopamine,n-arachidonyl dopamine,n-5z,8z,11z,14z-eicosatetraenoyl-dopamine,5z,8z,11z,14z-n-2-3,4-dihydroxyphenyl ethyl icosa-5,8,11,14-tetraenamide,n-2-3,4-dihydroxyphenyl ethyl-5z,8z,11z,14z-eicosatetraenamide
5282105
CHEBI:31231
(5Z,8Z,11Z,14Z)-N-[2-(3,4-dihydroxyphenyl)ethyl]icosa-5,8,11,14-tetraenamide
CCCCCC=CCC=CCC=CCC=CCCCC(=O)NCCC1=CC(=C(C=C1)O)O
Sensitivity Air Sensitive
Quantity 25mg
Formula Weight 439.64
Physical Form Viscous Oil
Percent Purity 98%
Chemical Name or Material N-Arachidonoyldopamine

storageNote1 : -30° to -10°C