Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-ArachidonylGABA, Tocris Bioscience™

Click to view available options
Quantity:
5mg
25mg
This item is not returnable. View return policy
128201-89-8
C24H39NO3
389.58
JKUDIEXTAYKJNX-CGRWFSSPSA-N
4-[[(5Z,8Z,11Z,14Z)-1-Oxo-5,8,11,14-eicosatetraenyl]amino]butanoic acid
44634744
4-[[(5E,8E,11E,14E)-icosa-5,8,11,14-tetraenoyl]amino]butanoic acid
CCCCCC=CCC=CCC=CCC=CCCCC(=O)NCCCC(=O)O
This item is not returnable. View return policy
128201-89-8
C24H39NO3
389.58
JKUDIEXTAYKJNX-CGRWFSSPSA-N
4-[[(5Z,8Z,11Z,14Z)-1-Oxo-5,8,11,14-eicosatetraenyl]amino]butanoic acid
44634744
4-[[(5E,8E,11E,14E)-icosa-5,8,11,14-tetraenoyl]amino]butanoic acid
CCCCCC=CCC=CCC=CCC=CCCCC(=O)NCCCC(=O)O

Inhibits pain in vivo

Arachidonyl amino acid; first isolated from bovine brain. Inhibits pain in vivo.
Quantity 25mg
Formula Weight 389.58
Percent Purity >98%
Chemical Name or Material N-ArachidonylGABA

Recommended Storage : Desiccate at -20°C