N-ArachidonylGABA, Tocris

Inhibits pain in vivo

Manufacturer: R&D Systems 1814/5

Quantity 5mg

 View more versions of this product

Catalog No. 18-145

  Check Availability
Add to Cart

    Chemical Identifiers

    Assay Percent Range >98%
    Molecular Formula C24H39NO3
    Formula Weight 389.58
    Synonym 4-[[(5Z,8Z,11Z,14Z)-1-Oxo-5,8,11,14-eicosatetraenyl]amino]butanoic acid
    Chemical Name or Material N-ArachidonylGABA

    Specifications

    Formula Weight 389.58
    Quantity 5mg
    Synonym 4-[[(5Z,8Z,11Z,14Z)-1-Oxo-5,8,11,14-eicosatetraenyl]amino]butanoic acid
    Assay Percent Range >98%
    Molecular Formula C24H39NO3
    CAS 128201-89-8
    Recommended Storage Desiccate at -20°C
    Chemical Name or Material N-ArachidonylGABA

    Desiccate at -20°C

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit