Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
N-Arachidonylglycine, Tocris Bioscience™
Click to view available options
Quantity:
5 mg
25 mg
Description
GPR18 agonist (EC50 = 44.5 nM). Endogenous anandamide-like compound. Lacks affinity for CB1 receptors (Ki > 10 μM), VR1 receptors (EC50 > 10 μM) and anandamide transporters (IC50 > 50 μM) but causes hot-plate analgesia in mice when given orally, and suppresses tonic inflammatory pain. Also endogenous GlyT2 inhibitor.
Specifications
Formula Weight | 361.52 |
Percent Purity | >98% |
Quantity | 25 mg |
Chemical Name or Material | N-Arachidonylglycine |
Safety and Handling
Recommended Storage | Desiccate at -20°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction