Alfa Aesar™ N-Benzoyl-2'-deoxyadenosine, 98+%

Manufacturer: Alfa Aesar™ H52303MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH52303MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 4546-72-9
    Molecular Formula C17H17N5O4
    Formula Weight 355.35
    MDL Number MFCD00009628
    Chemical Name or Material N-Benzoyl-2'-deoxyadenosine

    Specifications

    Formula Weight 355.35
    MDL Number MFCD00009628
    Quantity 250mg
    Molecular Formula C17H17N5O4
    CAS 4546-72-9
    Chemical Name or Material N-Benzoyl-2'-deoxyadenosine
    Assay ≥98%

    Description

    Applications
    Reactant in the synthesis of 2 ft., 5 ft.-Dideoxycytidines and Other Derivatives of 2 ft.-Deoxycytidine. Reactant in the synthesis of Mono- and Diamino Analogues of 2 ft.-Deoxyadenosine, Cordycepin, 9-(3-Deoxy-α-D-Threo-Pentofuranosyl)-Adenine and 9-(2-Deoxy-α-D-Threo-Pentofuranosyl)Adenine.

    Solubility
    Soluble in water (slightly), and methanol (20 mg/ml).

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit