Alfa Aesar™ N-Benzoyl-5'-O-tert-butyldimethylsilyl-2'-deoxyadenosine, 98+%

Manufacturer: Alfa Aesar™ H52309MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH52309MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 51549-39-4
    Molecular Formula C23H31N5O4Si
    Formula Weight 469.62
    MDL Number MFCD04972281
    Chemical Name or Material N-Benzoyl-5'-O-tert-butyldimethylsilyl-2'-deoxyadenosine

    Specifications

    Formula Weight 469.62
    Molecular Formula C23H31N5O4Si
    MDL Number MFCD04972281
    CAS 51549-39-4
    Quantity 250mg
    Chemical Name or Material N-Benzoyl-5'-O-tert-butyldimethylsilyl-2'-deoxyadenosine
    Assay ≥98%

    Description

    Applications
    N-Benzoyl-5 ft.-O-tert-butyldimethylsilyl-2 ft.-deoxyadenosine is used as pharmaceutical intermediate.

    Notes
    Stable under recommended storage conditions. Incompatible with oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit