Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Benzoyl-L-alanine, 95%

Click to view available options
Quantity:
5g
25g
100g
2198-64-3
C10H11NO3
193.202
MFCD00063128
UAQVHNZEONHPQG-ZETCQYMHSA-N
n-benzoyl-l-alanine,n-benzoylalanine,bz-ala-oh,l-alanine, n-benzoyl,s-alpha-methylhippuric acid,2s-2-phenylformamido propanoic acid,2s-2-benzamidopropanoic acid,2-benzoylamino-propionic acid,n-alpha-benzoyl-l-alanine,benzoylalanin
709778
CHEBI:71166
(2S)-2-benzamidopropanoic acid
CC(C(=O)O)NC(=O)C1=CC=CC=C1
2198-64-3
C10H11NO3
193.202
MFCD00063128
UAQVHNZEONHPQG-ZETCQYMHSA-N
n-benzoyl-l-alanine,n-benzoylalanine,bz-ala-oh,l-alanine, n-benzoyl,s-alpha-methylhippuric acid,2s-2-phenylformamido propanoic acid,2s-2-benzamidopropanoic acid,2-benzoylamino-propionic acid,n-alpha-benzoyl-l-alanine,benzoylalanin
709778
CHEBI:71166
(2S)-2-benzamidopropanoic acid
CC(C(=O)O)NC(=O)C1=CC=CC=C1
Formula Weight 193.2
Physical Form Solid
Percent Purity 95%
Quantity 25g
Chemical Name or Material N-Benzoyl-L-alanine

storageNote1 : -30° to -10°C