Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Benzoyl-L-glutamic acid, 98%

Click to view available options
Quantity:
5g
25g
100g
6094-36-6
C12H13NO5
251.238
MFCD00014028
LPJXPACOXRZCCP-VIFPVBQESA-N
bz-glu-oh,n-benzoyl-l-glutamic acid,benzoyl-l-glutamic acid,s-2-benzamidopentanedioic acid,n-benzoylglutamic acid,unii-3591d74mxp,2s-2-phenylformamido pentanedioic acid,+-n-benzoylglutamic acid,n-4-benzoyl-l-glutamic acid,2s-2-phenylcarbonylamino pentanedioic acid
11345724
(2S)-2-benzamidopentanedioic acid
C1=CC=C(C=C1)C(=O)NC(CCC(=O)O)C(=O)O
6094-36-6
C12H13NO5
251.238
MFCD00014028
LPJXPACOXRZCCP-VIFPVBQESA-N
bz-glu-oh,n-benzoyl-l-glutamic acid,benzoyl-l-glutamic acid,s-2-benzamidopentanedioic acid,n-benzoylglutamic acid,unii-3591d74mxp,2s-2-phenylformamido pentanedioic acid,+-n-benzoylglutamic acid,n-4-benzoyl-l-glutamic acid,2s-2-phenylcarbonylamino pentanedioic acid
11345724
(2S)-2-benzamidopentanedioic acid
C1=CC=C(C=C1)C(=O)NC(CCC(=O)O)C(=O)O
Beilstein 2622989
Formula Weight 251.24
Physical Form Solid
Percent Purity 98%
Quantity 100g
Chemical Name or Material N-Benzoyl-L-glutamic acid