Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Benzoyl-L-phenylalanine, 98%

Click to view available options
Quantity:
5g
25g
2566-22-5
C16H15NO3
269.3
MFCD00063153
NPKISZUVEBESJI-AWEZNQCLSA-N
n-benzoyl-l-phenylalanine,benzoylphenylalanine,bz-phe-oh,l-bpa,unii-9s304jl9m7,benzoyl-l-phenylalanine,l-phenylalanine, n-benzoyl,2s-3-phenyl-2-phenylformamido propanoic acid,2s-2-benzoylamino-3-phenylpropanoic acid,n-benzoylphenylalanine #
97370
(2S)-2-benzamido-3-phenylpropanoic acid
C1=CC=C(C=C1)CC(C(=O)O)NC(=O)C2=CC=CC=C2
2566-22-5
C16H15NO3
269.3
MFCD00063153
NPKISZUVEBESJI-AWEZNQCLSA-N
n-benzoyl-l-phenylalanine,benzoylphenylalanine,bz-phe-oh,l-bpa,unii-9s304jl9m7,benzoyl-l-phenylalanine,l-phenylalanine, n-benzoyl,2s-3-phenyl-2-phenylformamido propanoic acid,2s-2-benzoylamino-3-phenylpropanoic acid,n-benzoylphenylalanine #
97370
(2S)-2-benzamido-3-phenylpropanoic acid
C1=CC=C(C=C1)CC(C(=O)O)NC(=O)C2=CC=CC=C2
Formula Weight 269.3
Physical Form Powder
Percent Purity 98%
Color White
Quantity 5g
Chemical Name or Material N-Benzoyl-L-phenylalanine

RTECSNumber : SQ7326700

storageNote1 : 2° to 8°C