Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Benzyl-2-pyridineethylamine, Thermo Scientific™

Catalog Number AAH5025703
Click to view available options
Quantity:
250 mg
1 g
6312-25-0
C14H16N2
212.296
MFCD00089263
WUDUNBALAGWKQB-UHFFFAOYSA-N
n-benzyl-2-pyridin-2-yl ethanamine, n-benzyl-2-pyridin-2-yl-ethanamine, benzyl 2-pyridin-2-yl ethyl amine, n-benzyl-2-pyridin-2-ylethylamine, benzyl 2-2-pyridyl ethyl amine, acmc-20ak6u, n-benzyl-2-pyridineethylamine, 2-2-benzylaminoethyl pyridine, benzyl-2-pyridin-2-yl-ethyl amine, n-phenylmethyl-2-pyridin-2-ylethanamine
238354
N-benzyl-2-pyridin-2-ylethanamine
C1=CC=C(C=C1)CNCCC2=CC=CC=N2
6312-25-0
C14H16N2
212.296
MFCD00089263
WUDUNBALAGWKQB-UHFFFAOYSA-N
n-benzyl-2-pyridin-2-yl ethanamine, n-benzyl-2-pyridin-2-yl-ethanamine, benzyl 2-pyridin-2-yl ethyl amine, n-benzyl-2-pyridin-2-ylethylamine, benzyl 2-2-pyridyl ethyl amine, acmc-20ak6u, n-benzyl-2-pyridineethylamine, 2-2-benzylaminoethyl pyridine, benzyl-2-pyridin-2-yl-ethyl amine, n-phenylmethyl-2-pyridin-2-ylethanamine
238354
N-benzyl-2-pyridin-2-ylethanamine
C1=CC=C(C=C1)CNCCC2=CC=CC=N2

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
UN Number UN2735
Formula Weight 212.3
Chemical Name or Material N-Benzyl-2-pyridineethylamine
Hazard Category Danger
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit