Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Benzyl-N-ethyl-3-methylaniline, 98%

Click to view available options
Quantity:
250mg
1g
5g
119-94-8
C16H19N
225.335
MFCD00025950
VIACAIAQHPLINF-UHFFFAOYSA-N
n-benzyl-n-ethyl-m-toluidine,n-ethyl-n-benzyl-m-toluidine,ethylbenzyltoluidine,benzenemethanamine, n-ethyl-n-3-methylphenyl,3-n-benzyl-n-ethylamino toluene,n-ethyl-n-benzyl-meta-toluidine,3-n-benzyl-n-ethylamino methylbenzene,m-toluidine, n-benzyl-n-ethyl,n-benzyl-n-ethyl-meta-toluidine,n-ethyl-n-m-tolyl benzylamine
67076
N-benzyl-N-ethyl-3-methylaniline
CCN(CC1=CC=CC=C1)C2=CC=CC(=C2)C
119-94-8
C16H19N
225.335
MFCD00025950
VIACAIAQHPLINF-UHFFFAOYSA-N
n-benzyl-n-ethyl-m-toluidine,n-ethyl-n-benzyl-m-toluidine,ethylbenzyltoluidine,benzenemethanamine, n-ethyl-n-3-methylphenyl,3-n-benzyl-n-ethylamino toluene,n-ethyl-n-benzyl-meta-toluidine,3-n-benzyl-n-ethylamino methylbenzene,m-toluidine, n-benzyl-n-ethyl,n-benzyl-n-ethyl-meta-toluidine,n-ethyl-n-m-tolyl benzylamine
67076
N-benzyl-N-ethyl-3-methylaniline
CCN(CC1=CC=CC=C1)C2=CC=CC(=C2)C
Quantity 250mg
Density 0.77g/mL
Boiling Point 161°C to 162°C
UN Number UN2753
Beilstein 5336067
Formula Weight 225.33
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material N-Benzyl-N-ethyl-3-methylaniline

GHS H Statement
H301-H311-H331-H227
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Combustible liquid.

GHS P Statement
P210-P261-P301+P310-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: N-ETHYLBENZYLTOLUIDINES LIQUID

TSCA : TSCA