Alfa Aesar™ N-Benzylbenzamide, 99+%

Manufacturer: Alfa Aesar™ A1220414

Quantity 25g

 View more versions of this product

Catalog No. AAA1220414

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 1485-70-7
    Molecular Formula C6H5CONHCH2C6H5
    Formula Weight 211.26
    MDL Number MFCD00003070
    Beilstein 2211315
    Synonym N-Benzoylbenzylamine
    Chemical Name or Material N-Benzylbenzamide

    Specifications

    Formula Weight 211.26
    MDL Number MFCD00003070
    CAS 1485-70-7
    Beilstein 2211315
    Synonym N-Benzoylbenzylamine
    Molecular Formula C6H5CONHCH2C6H5
    Melting Point 104° to 108°C
    Quantity 25g
    Chemical Name or Material N-Benzylbenzamide
    Assay ≥99%

    Description

    Applications
    N-Benzylbenzamide is a convenient precursor to α-substituted benzyl amines and an indicator for the titration of butyl lithium and other lithium bases.

    Solubility
    Soluble in Acetone (25 mg/ml).

    Notes
    Store in cool, dry place in a tightly sealed container. Store away from oxidizing agent.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit