Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Benzylbenzamide, 99+%

Manufacturer:  Alfa Aesar™ A1220422

Quantity 100g

 View more versions of this product

Catalog No. AAA1220422

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Description

Applications
N-Benzylbenzamide is a convenient precursor to α-substituted benzyl amines and an indicator for the titration of butyl lithium and other lithium bases.

Solubility
Soluble in Acetone (25 mg/ml).

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agent.
Description & Specifications

Chemical Identifiers

1485-70-7
211.26
2211315
N-Benzylbenzamide
C6H5CONHCH2C6H5
MFCD00003070
N-Benzoylbenzylamine

Specifications

1485-70-7
211.26
MFCD00003070
100g
N-Benzylbenzamide
C6H5CONHCH2C6H5
104° to 108°C
2211315
N-Benzoylbenzylamine
≥99%
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit