Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Benzylbenzamide, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
Plastic bottle
1485-70-7
C14H13NO
211.26
MFCD00003070
LKQUCICFTHBFAL-UHFFFAOYSA-N
benzamide, n-phenylmethyl,n-benzyl-benzamide,phenyl-n-benzylcarboxamide,benzoylbenzylamine,n~1~-benzylbenzamide,benzamide, n-benzyl,n-phenylmethyl benzamide,acmc-1bx9y,n-benzylbenzamide,lkqucicfthbfal-uhfffaoysa
73878
N-benzylbenzamide
C1=CC=C(C=C1)CNC(=O)C2=CC=CC=C2
1485-70-7
C14H13NO
211.26
MFCD00003070
LKQUCICFTHBFAL-UHFFFAOYSA-N
benzamide, n-phenylmethyl,n-benzyl-benzamide,phenyl-n-benzylcarboxamide,benzoylbenzylamine,n~1~-benzylbenzamide,benzamide, n-benzyl,n-phenylmethyl benzamide,acmc-1bx9y,n-benzylbenzamide,lkqucicfthbfal-uhfffaoysa
73878
N-benzylbenzamide
C1=CC=C(C=C1)CNC(=O)C2=CC=CC=C2
Quantity 5g
Color White
Infrared Spectrum Authentic
Packaging Glass bottle
Melting Point 104.0°C to 107.0°C
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (HPLC)
Linear Formula C6H5CONHCH2C6H5
Solubility Information Solubility in water: insoluble
Formula Weight 211.26
Physical Form Fine Crystalline Powder
Percent Purity 99%
Chemical Name or Material N-Benzylbenzamide, 99%
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face prot

GHS Signal Word: Warning

EINECSNumber : 216-063-4