Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Benzylformamide, 99%

Click to view available options
Quantity:
5g
25g
100g
6343-54-0
C8H9NO
135.166
MFCD00003281
IIBOGKHTXBPGEI-UHFFFAOYSA-N
benzyl formamide,n-phenylmethyl formamide,n-benzyl-formamide,unii-l363d92elk,formamide, n-phenylmethyl,formamide,n-phenylmethyl,chembl46293,n-benzyl formamide,n-benzylcarboxamide,n-formylbenzylamine
80654
CHEBI:41117
N-benzylformamide
C1=CC=C(C=C1)CNC=O
6343-54-0
C8H9NO
135.166
MFCD00003281
IIBOGKHTXBPGEI-UHFFFAOYSA-N
benzyl formamide,n-phenylmethyl formamide,n-benzyl-formamide,unii-l363d92elk,formamide, n-phenylmethyl,formamide,n-phenylmethyl,chembl46293,n-benzyl formamide,n-benzylcarboxamide,n-formylbenzylamine
80654
CHEBI:41117
N-benzylformamide
C1=CC=C(C=C1)CNC=O
Melting Point 60°C to 61°C
Quantity 100g
Formula Weight 135.17
Percent Purity 99%
Chemical Name or Material N-Benzylformamide