Alfa Aesar™ N-Benzylformamide, 99%

Manufacturer: Alfa Aesar™ B2372206

Quantity 5g

 View more versions of this product

Catalog No. AAB2372206

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 6343-54-0
    Molecular Formula C6H5CH2NHOCH
    Formula Weight 135.17
    MDL Number MFCD00003281
    Synonym N-Formylbenzylamine
    Chemical Name or Material N-Benzylformamide

    Specifications

    Formula Weight 135.17
    Molecular Formula C6H5CH2NHOCH
    Melting Point 60° to 61°C
    MDL Number MFCD00003281
    CAS 6343-54-0
    Quantity 5g
    Synonym N-Formylbenzylamine
    Chemical Name or Material N-Benzylformamide
    Assay 99%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit