Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Benzylidenebenzylamine, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
780-25-6
C14H13N
195.265
MIYKHJXFICMPOJ-UHFFFAOYSA-N
n-benzylidenebenzylamine,benzylidenebenzylamine,benzenemethanamine, n-phenylmethylene,n-phenylmethylene benzenemethanamine,benzaldehyde n-benzylimine,benzylamine, n-benzylidene,n-benzylaminebenzaldehyde,n-benzylidene-1-phenylmethanamine,e-n-benzylidene-1-phenylmethanamine,benzyl phenylmethylidene amine
95831
N-benzyl-1-phenylmethanimine
C1=CC=C(C=C1)CN=CC2=CC=CC=C2
780-25-6
C14H13N
195.265
MIYKHJXFICMPOJ-UHFFFAOYSA-N
n-benzylidenebenzylamine,benzylidenebenzylamine,benzenemethanamine, n-phenylmethylene,n-phenylmethylene benzenemethanamine,benzaldehyde n-benzylimine,benzylamine, n-benzylidene,n-benzylaminebenzaldehyde,n-benzylidene-1-phenylmethanamine,e-n-benzylidene-1-phenylmethanamine,benzyl phenylmethylidene amine
95831
N-benzyl-1-phenylmethanimine
C1=CC=C(C=C1)CN=CC2=CC=CC=C2
Infrared Spectrum Authentic
Quantity 25g
Color Colorless to Yellow
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Formula Weight 195.27
Physical Form Liquid
Percent Purity ≥98.5%
Chemical Name or Material N-Benzylidenebenzylamine

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.