Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ N-Benzylidenemethylamine, 98+%
Click to view available options
Quantity:
5g
25g
100g
Specifications
Density | 0.967 |
Refractive Index | 1.552 |
Flash Point | 77°C (170°F) |
Quantity | 100g |
Boiling Point | 66°C to 69°C (10mmHg) |
Beilstein | 878278 |
Formula Weight | 119.17 |
Percent Purity | ≥98% |
Chemical Name or Material | N-Benzylidenemethylamine |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 210-728-2
TSCA : Yes
Recommended Storage : Keep cold