Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Benzylmaleamic acid, 98+%

Click to view available options
Quantity:
5g
25g
100g
15329-69-8
C11H11NO3
205.213
MFCD00020482
BHWGQIYJCMMSNM-SREVYHEPSA-N
n-benzylmaleamic acid,z-4-benzylamino-4-oxobut-2-enoic acid,2z-3-benzylcarbamoyl prop-2-enoic acid,2e-4-benzylamino-4-oxobut-2-enoic acid,n-benzyl maleamic acid,maleic acid monobenzylamide,z-3-benzylcarbamoyl-acrylic acid,2z-3-n-benzylcarbamoyl prop-2-enoic acid,2z-4-benzylamino-4-oxobut-2-enoic acid
1549764
(Z)-4-(benzylamino)-4-oxobut-2-enoic acid
C1=CC=C(C=C1)CNC(=O)C=CC(=O)O
15329-69-8
C11H11NO3
205.213
MFCD00020482
BHWGQIYJCMMSNM-SREVYHEPSA-N
n-benzylmaleamic acid,z-4-benzylamino-4-oxobut-2-enoic acid,2z-3-benzylcarbamoyl prop-2-enoic acid,2e-4-benzylamino-4-oxobut-2-enoic acid,n-benzyl maleamic acid,maleic acid monobenzylamide,z-3-benzylcarbamoyl-acrylic acid,2z-3-n-benzylcarbamoyl prop-2-enoic acid,2z-4-benzylamino-4-oxobut-2-enoic acid
1549764
(Z)-4-(benzylamino)-4-oxobut-2-enoic acid
C1=CC=C(C=C1)CNC(=O)C=CC(=O)O
Quantity 5g
Melting Point 136°C to 138°C
Beilstein 1212733
Formula Weight 205.21
Percent Purity ≥98%
Chemical Name or Material N-Benzylmaleamic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning