Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Benzylmaleimide, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
1631-26-1
C11H9NO2
187.198
MFCD00014540
MKRBAPNEJMFMHU-UHFFFAOYSA-N
n-benzylmaleimide,1-benzyl-1h-pyrrole-2,5-dione,n-benzyl maleimide,1h-pyrrole-2,5-dione, 1-phenylmethyl,1-benzylmaleimide,n-benzylmaleinimide,1-benzyl-pyrrole-2,5-dione,n-benzyl-maleimide,1-benzyl-2,5-dihydro-1h-pyrrole-2,5-dione,n-benzylmaleimide dihydrate
74204
1-benzylpyrrole-2,5-dione
C1=CC=C(C=C1)CN2C(=O)C=CC2=O
1631-26-1
C11H9NO2
187.198
MFCD00014540
MKRBAPNEJMFMHU-UHFFFAOYSA-N
n-benzylmaleimide,1-benzyl-1h-pyrrole-2,5-dione,n-benzyl maleimide,1h-pyrrole-2,5-dione, 1-phenylmethyl,1-benzylmaleimide,n-benzylmaleinimide,1-benzyl-pyrrole-2,5-dione,n-benzyl-maleimide,1-benzyl-2,5-dihydro-1h-pyrrole-2,5-dione,n-benzylmaleimide dihydrate
74204
1-benzylpyrrole-2,5-dione
C1=CC=C(C=C1)CN2C(=O)C=CC2=O
Quantity 1g
Infrared Spectrum Authentic
Color White to Yellow
Packaging Glass bottle
Melting Point 68°C to 71°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 187.2
Physical Form Crystalline Powder or Needles
Percent Purity ≥97.5%
Chemical Name or Material N-Benzylmaleimide
Show More Show Less

GHS H Statement:
Toxic if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if inhaled.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger