Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Benzylnaltrindole hydrochloride, Tocris Bioscience™

Click to view available options
Quantity:
10mg
50mg
1206487-81-1
C33H33ClN2O3
541.088
KDEBSUYWJUBPCB-REBVNRAYSA-N
n-benzylnaltrindole hydrochloride
131845649
C1CC1CN2CCC34C5C6=C(CC3(C2CC7=C4C(=C(C=C7)O)O5)O)C8=CC=CC=C8N6CC9=CC=CC=C9.Cl
1206487-81-1
C33H33ClN2O3
541.088
KDEBSUYWJUBPCB-REBVNRAYSA-N
n-benzylnaltrindole hydrochloride
131845649
C1CC1CN2CCC34C5C6=C(CC3(C2CC7=C4C(=C(C=C7)O)O5)O)C8=CC=CC=C8N6CC9=CC=CC=C9.Cl

Selective non-peptide δ2 antagonist

Potent δ2-selective opioid receptor antagonist with a long duration of action in vivo.
Quantity 50mg
Formula Weight 541.09
Percent Purity >98%
Chemical Name or Material N-Benzylnaltrindole hydrochloride

Recommended Storage : Store at Room Temperature