Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Benzyloxycarbonyl-3-(Fmoc-amino)-L-alanine, 95%

Click to view available options
Quantity:
250mg
1g
5g
142855-80-9
MFCD00274016
Z-Dap(Fmoc)-OH
142855-80-9
MFCD00274016
Z-Dap(Fmoc)-OH
Beilstein 6356947
Formula Weight 460.49
Physical Form Powder
Percent Purity 95%
Color White
Quantity 1g
Chemical Name or Material N-Benzyloxycarbonyl-3-(Fmoc-amino)-L-alanine

storageNote1 : 2° to 8°C