Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-(Benzyloxycarbonyl)alanyl-L-alaninamide, 97%

Click to view available options
Quantity:
1g
5g
50444-54-7
C14H19N3O4
293.323
MFCD00238398
WEIOJLPDGBBVCH-UWVGGRQHSA-N
z-ala-ala-nh2,benzyl s-1-s-1-amino-1-oxopropan-2-yl amino-1-oxopropan-2-yl carbamate,benzyl n-1s-1-1s-1-carbamoylethyl carbamoyl ethyl carbamate,z-l-ala-l-ala-nh2,benzyl n-2s-1-2s-1-amino-1-oxopropan-2-yl amino-1-oxopropan-2-yl carbamate,benzyl s-1-s-1-amino-1-oxopropan-2-yl amino-1-oxopropan-2-yl carbamate
6993440
benzyl N-[(2S)-1-[[(2S)-1-amino-1-oxopropan-2-yl]amino]-1-oxopropan-2-yl]carbamate
CC(C(=O)N)NC(=O)C(C)NC(=O)OCC1=CC=CC=C1
50444-54-7
C14H19N3O4
293.323
MFCD00238398
WEIOJLPDGBBVCH-UWVGGRQHSA-N
z-ala-ala-nh2,benzyl s-1-s-1-amino-1-oxopropan-2-yl amino-1-oxopropan-2-yl carbamate,benzyl n-1s-1-1s-1-carbamoylethyl carbamoyl ethyl carbamate,z-l-ala-l-ala-nh2,benzyl n-2s-1-2s-1-amino-1-oxopropan-2-yl amino-1-oxopropan-2-yl carbamate,benzyl s-1-s-1-amino-1-oxopropan-2-yl amino-1-oxopropan-2-yl carbamate
6993440
benzyl N-[(2S)-1-[[(2S)-1-amino-1-oxopropan-2-yl]amino]-1-oxopropan-2-yl]carbamate
CC(C(=O)N)NC(=O)C(C)NC(=O)OCC1=CC=CC=C1
Formula Weight 293.32
Physical Form Solid
Percent Purity 97%
Quantity 1g
Chemical Name or Material N-(Benzyloxycarbonyl)alanyl-L-alaninamide

storageNote1 : 2° to 8°C