Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Benzyloxycarbonyl-D-alanylglycine, 95%

Click to view available options
Quantity:
250mg
1g
5g
34286-66-3
C13H16N2O5
280.28
MFCD00238424
RNBMQRYMCAVZPN-SECBINFHSA-N
z-d-ala-gly-oh,r-2-2-benzyloxy carbonyl amino propanamido acetic acid,n-r-2-benzyloxycarbonyl amino-1-oxopropyl glycine,r-2-2-benzyloxy carbonyl amino propanamido aceticacid,2-2r-2-benzyloxy carbonyl amino propanamido acetic acid,2r-2-benzyloxy carbonyl amino propanamido acetic acid,2-2r-2-phenylmethoxycarbonylamino propanoyl amino acetic acid
7004845
2-[[(2R)-2-(phenylmethoxycarbonylamino)propanoyl]amino]acetic acid
CC(C(=O)NCC(=O)O)NC(=O)OCC1=CC=CC=C1
34286-66-3
C13H16N2O5
280.28
MFCD00238424
RNBMQRYMCAVZPN-SECBINFHSA-N
z-d-ala-gly-oh,r-2-2-benzyloxy carbonyl amino propanamido acetic acid,n-r-2-benzyloxycarbonyl amino-1-oxopropyl glycine,r-2-2-benzyloxy carbonyl amino propanamido aceticacid,2-2r-2-benzyloxy carbonyl amino propanamido acetic acid,2r-2-benzyloxy carbonyl amino propanamido acetic acid,2-2r-2-phenylmethoxycarbonylamino propanoyl amino acetic acid
7004845
2-[[(2R)-2-(phenylmethoxycarbonylamino)propanoyl]amino]acetic acid
CC(C(=O)NCC(=O)O)NC(=O)OCC1=CC=CC=C1
Formula Weight 280.28
Physical Form Solid
Percent Purity 95%
Quantity 1g
Chemical Name or Material N-Benzyloxycarbonyl-D-alanylglycine

storageNote1 : 2° to 8°C