Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Benzyloxycarbonyl-D-aspartic acid 4-benzyl ester, 95%

Click to view available options
Quantity:
250mg
1g
5g
5241-62-3
C19H19NO6
357.362
MFCD00237336
VUKCNAATVIWRTF-MRXNPFEDSA-N
z-d-asp obzl-oh,z-d-asp-obzl,2r-4-benzyloxy-2-benzyloxy carbonyl amino-4-oxobutanoic acid,2r-4-benzyloxy-2-benzyloxy carbonyl amino-4-oxobutanoic acid,n-benzyloxycarbonyl-d-aspartic acid 4-benzyl ester,2r-4-oxo-4-phenylmethoxy-2-phenylmethoxycarbonylamino butanoic acid,2r-4-benzyloxy-2-benzyloxy carbonyl amino-4-oxobutanoic acid non-preferred name,2r-4-benzyloxy-2-benzyloxy carbonyl amino-4-oxobutanoic acid non-preferred name
47604
(2R)-4-oxo-4-phenylmethoxy-2-(phenylmethoxycarbonylamino)butanoic acid
C1=CC=C(C=C1)COC(=O)CC(C(=O)O)NC(=O)OCC2=CC=CC=C2
5241-62-3
C19H19NO6
357.362
MFCD00237336
VUKCNAATVIWRTF-MRXNPFEDSA-N
z-d-asp obzl-oh,z-d-asp-obzl,2r-4-benzyloxy-2-benzyloxy carbonyl amino-4-oxobutanoic acid,2r-4-benzyloxy-2-benzyloxy carbonyl amino-4-oxobutanoic acid,n-benzyloxycarbonyl-d-aspartic acid 4-benzyl ester,2r-4-oxo-4-phenylmethoxy-2-phenylmethoxycarbonylamino butanoic acid,2r-4-benzyloxy-2-benzyloxy carbonyl amino-4-oxobutanoic acid non-preferred name,2r-4-benzyloxy-2-benzyloxy carbonyl amino-4-oxobutanoic acid non-preferred name
47604
(2R)-4-oxo-4-phenylmethoxy-2-(phenylmethoxycarbonylamino)butanoic acid
C1=CC=C(C=C1)COC(=O)CC(C(=O)O)NC(=O)OCC2=CC=CC=C2
Beilstein 2570779
Formula Weight 357.36
Physical Form Solid
Percent Purity 95%
Melting Point 105°C to 109°C
Quantity 1g
Chemical Name or Material N-Benzyloxycarbonyl-D-aspartic acid 4-benzyl ester

storageNote1 : 2° to 8°C