Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Benzyloxycarbonyl-D-norvaline, 95%

Click to view available options
Quantity:
5g
25g
42918-89-8
C13H17NO4
251.282
MFCD00136826
NSJDRLWFFAWSFP-LLVKDONJSA-N
z-d-nva-oh,n-cbz-d-norvaline,cbz-d-norvaline,z-d-nva-oh,z-cbz-d-norvaline,n-benzyloxycarbonyl-d-norvaline,n-benzyloxycarbonyl-d-norvaline,2r-2-benzyloxy carbonyl amino pentanoic acid,r-2-benzyloxycarbonylamino pentanoic acid,d-norvaline,n-phenylmethoxy carbonyl
10106192
(2R)-2-(phenylmethoxycarbonylamino)pentanoic acid
CCCC(C(=O)O)NC(=O)OCC1=CC=CC=C1
42918-89-8
C13H17NO4
251.282
MFCD00136826
NSJDRLWFFAWSFP-LLVKDONJSA-N
z-d-nva-oh,n-cbz-d-norvaline,cbz-d-norvaline,z-d-nva-oh,z-cbz-d-norvaline,n-benzyloxycarbonyl-d-norvaline,n-benzyloxycarbonyl-d-norvaline,2r-2-benzyloxy carbonyl amino pentanoic acid,r-2-benzyloxycarbonylamino pentanoic acid,d-norvaline,n-phenylmethoxy carbonyl
10106192
(2R)-2-(phenylmethoxycarbonylamino)pentanoic acid
CCCC(C(=O)O)NC(=O)OCC1=CC=CC=C1
Beilstein 2466156
Formula Weight 251.48
Physical Form Powder
Percent Purity 95%
Color White
Melting Point 84°C to 90°C
Quantity 25g
Chemical Name or Material N-Benzyloxycarbonyl-D-norvaline

storageNote1 : -30° to -10°C