Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Benzyloxycarbonyl-D-phenylalaninamide, 97%

Click to view available options
Quantity:
1g
5g
5241-56-5
C17H18N2O3
298.342
MFCD00191136
HHBOFAIEPRHUSR-OAHLLOKOSA-N
r-benzyl 1-amino-1-oxo-3-phenylpropan-2-yl carbamate,z-d-phe-nh2,benzyl n-1r-1-carbamoyl-2-phenylethyl carbamate,nalpha-benzyloxy carbonyl-d-phenylalaninamide,r-benzyl 1-amino-1-oxo-3-phenylpropan-2-ylcarbamate
68077307
benzyl N-[(2R)-1-amino-1-oxo-3-phenylpropan-2-yl]carbamate
C1=CC=C(C=C1)CC(C(=O)N)NC(=O)OCC2=CC=CC=C2
5241-56-5
C17H18N2O3
298.342
MFCD00191136
HHBOFAIEPRHUSR-OAHLLOKOSA-N
r-benzyl 1-amino-1-oxo-3-phenylpropan-2-yl carbamate,z-d-phe-nh2,benzyl n-1r-1-carbamoyl-2-phenylethyl carbamate,nalpha-benzyloxy carbonyl-d-phenylalaninamide,r-benzyl 1-amino-1-oxo-3-phenylpropan-2-ylcarbamate
68077307
benzyl N-[(2R)-1-amino-1-oxo-3-phenylpropan-2-yl]carbamate
C1=CC=C(C=C1)CC(C(=O)N)NC(=O)OCC2=CC=CC=C2
Quantity 5g
Formula Weight 298.34
Physical Form Solid
Percent Purity 97%
Chemical Name or Material N-Benzyloxycarbonyl-D-phenylalaninamide

storageNote1 : 2° to 8°C