Alfa Aesar™ N-Benzyloxycarbonyl-D-phenylalaninol, 98%

Manufacturer: Alfa Aesar™ H2781914

Quantity 25g

 View more versions of this product

Catalog No. AAH2781914

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 58917-85-4
    Molecular Formula C17H19NO3
    Formula Weight 285.35
    MDL Number MFCD00191193
    Synonym (R)-2-Carbobenzyloxyamino-3-phenyl-1-propanol; Z-D-Phe-ol
    Chemical Name or Material N-Benzyloxycarbonyl-D-phenylalaninol

    Specifications

    Formula Weight 285.35
    Melting Point 92° to 95°C
    Quantity 25g
    Molecular Formula C17H19NO3
    MDL Number MFCD00191193
    CAS 58917-85-4
    Synonym (R)-2-Carbobenzyloxyamino-3-phenyl-1-propanol; Z-D-Phe-ol
    Optical Rotation +42° (c=1 in Methanol)
    Chemical Name or Material N-Benzyloxycarbonyl-D-phenylalaninol
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit