Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Benzyloxycarbonyl-D-proline, 98+%

Click to view available options
Quantity:
1g
5g
25g
6404-31-5
C13H15NO4
249.266
MFCD00063228
JXGVXCZADZNAMJ-LLVKDONJSA-N
n-benzyloxycarbonyl-d-proline,z-d-pro-oh,+-z-d-proline,z-d-proline,r-n-benzyloxycarbonylproline,n-cbz-d-proline,n-carbobenzoxy-d-proline,1-benzyloxy carbonyl-d-proline,r-1-benzyloxy carbonyl pyrrolidine-2-carboxylic acid,2r-1-benzyloxy carbonyl pyrrolidine-2-carboxylic acid
111208
(2R)-1-phenylmethoxycarbonylpyrrolidine-2-carboxylic acid
C1CC(N(C1)C(=O)OCC2=CC=CC=C2)C(=O)O
6404-31-5
C13H15NO4
249.266
MFCD00063228
JXGVXCZADZNAMJ-LLVKDONJSA-N
n-benzyloxycarbonyl-d-proline,z-d-pro-oh,+-z-d-proline,z-d-proline,r-n-benzyloxycarbonylproline,n-cbz-d-proline,n-carbobenzoxy-d-proline,1-benzyloxy carbonyl-d-proline,r-1-benzyloxy carbonyl pyrrolidine-2-carboxylic acid,2r-1-benzyloxy carbonyl pyrrolidine-2-carboxylic acid
111208
(2R)-1-phenylmethoxycarbonylpyrrolidine-2-carboxylic acid
C1CC(N(C1)C(=O)OCC2=CC=CC=C2)C(=O)O
Quantity 1g
Melting Point 76°C to 78°C
Optical Rotation +40° (c=2 in Ethanol)
Beilstein 485188
Formula Weight 249.27
Percent Purity ≥98%
Chemical Name or Material N-Benzyloxycarbonyl-D-proline

Recommended Storage : 2° to 8°C