Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Benzyloxycarbonyl-DL-asparagine, 98%

Click to view available options
Quantity:
5g
25g
100g
29880-22-6
C12H14N2O5
266.253
MFCD00065101
FUCKRCGERFLLHP-UHFFFAOYSA-N
z-dl-asparagine,z-dl-asn-oh,n-carbobenzyloxy-d-asparagine,n-carbobenzoxy-dl-asparagine,2-benzyloxy carbonyl amino-3-carbamoylpropanoic acid,nalpha-carbobenzoxy-dl-asparagine,carbobenzoxy-l-asparagine,4-amino-4-oxo-2-phenylmethoxycarbonylamino butanoic acid,asparagine, n 2-benzyl ester, l,benzyloxycarbonyl-asparagine
246565
4-amino-4-oxo-2-(phenylmethoxycarbonylamino)butanoic acid
C1=CC=C(C=C1)COC(=O)NC(CC(=O)N)C(=O)O
29880-22-6
C12H14N2O5
266.253
MFCD00065101
FUCKRCGERFLLHP-UHFFFAOYSA-N
z-dl-asparagine,z-dl-asn-oh,n-carbobenzyloxy-d-asparagine,n-carbobenzoxy-dl-asparagine,2-benzyloxy carbonyl amino-3-carbamoylpropanoic acid,nalpha-carbobenzoxy-dl-asparagine,carbobenzoxy-l-asparagine,4-amino-4-oxo-2-phenylmethoxycarbonylamino butanoic acid,asparagine, n 2-benzyl ester, l,benzyloxycarbonyl-asparagine
246565
4-amino-4-oxo-2-(phenylmethoxycarbonylamino)butanoic acid
C1=CC=C(C=C1)COC(=O)NC(CC(=O)N)C(=O)O
Beilstein 6066596
Formula Weight 266.25
Physical Form Powder
Percent Purity 98%
Color White
Quantity 25g
Chemical Name or Material N-Benzyloxycarbonyl-DL-asparagine