Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Benzyloxycarbonyl-DL-aspartic acid, 95%

Click to view available options
Quantity:
5g
25g
100g
4515-21-3
C12H13NO6
267.237
MFCD00063181
XYXYXSKSTZAEJW-UHFFFAOYSA-N
z-dl-asp-oh,n-carbobenzoxy-dl-aspartic acid,z-dl-aspartic acid,cbz-dl-aspartic acid,2-benzyloxy carbonyl amino succinic acid,n-cbz-dl-aspartic acid,n-benzyloxy carbonyl aspartic acid,2-benzyloxycarbonylamino butanedioic acid,2-phenylmethoxycarbonylamino butanedioic acid,2-benzyloxy carbonyl amino butanedioic acid
70855
2-(phenylmethoxycarbonylamino)butanedioic acid
C1=CC=C(C=C1)COC(=O)NC(CC(=O)O)C(=O)O
4515-21-3
C12H13NO6
267.237
MFCD00063181
XYXYXSKSTZAEJW-UHFFFAOYSA-N
z-dl-asp-oh,n-carbobenzoxy-dl-aspartic acid,z-dl-aspartic acid,cbz-dl-aspartic acid,2-benzyloxy carbonyl amino succinic acid,n-cbz-dl-aspartic acid,n-benzyloxy carbonyl aspartic acid,2-benzyloxycarbonylamino butanedioic acid,2-phenylmethoxycarbonylamino butanedioic acid,2-benzyloxy carbonyl amino butanedioic acid
70855
2-(phenylmethoxycarbonylamino)butanedioic acid
C1=CC=C(C=C1)COC(=O)NC(CC(=O)O)C(=O)O
Beilstein 4701629
Formula Weight 267.24
Physical Form Powder
Percent Purity 95%
Color White
Quantity 5g
Chemical Name or Material N-Benzyloxycarbonyl-DL-aspartic acid

storageNote1 : -30° to -10°C