Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Benzyloxycarbonyl-L-aspartic acid 4-tert-butyl ester 1-methyl ester, 98%

Click to view available options
Quantity:
5g
25g
63327-57-1
C17H23NO6
337.372
MFCD00270581
HHCORPWNIMSWJP-ZDUSSCGKSA-N
z-asp otbu-ome,s-4-tert-butyl 1-methyl 2-benzyloxy carbonyl amino succinate,z-l-aspartic acid beta-tert-butyl ester alpha-methyl ester,4-tert-butyl 1-methyl 2s-2-benzyloxy carbonyl amino butanedioate,z-l-asp otbu-ome,4-tert-butyl 1-methyl n-benzyloxy carbonyl-l-aspartate,n-benzyloxycarbonyl-l-aspartic acid 1-methyl 4-tert-butyl ester,s-2-benzyloxycarbonylamino-succinic acid 4-tert-butyl ester 1-methyl ester
10593022
4-O-tert-butyl 1-O-methyl (2S)-2-(phenylmethoxycarbonylamino)butanedioate
CC(C)(C)OC(=O)CC(C(=O)OC)NC(=O)OCC1=CC=CC=C1
63327-57-1
C17H23NO6
337.372
MFCD00270581
HHCORPWNIMSWJP-ZDUSSCGKSA-N
z-asp otbu-ome,s-4-tert-butyl 1-methyl 2-benzyloxy carbonyl amino succinate,z-l-aspartic acid beta-tert-butyl ester alpha-methyl ester,4-tert-butyl 1-methyl 2s-2-benzyloxy carbonyl amino butanedioate,z-l-asp otbu-ome,4-tert-butyl 1-methyl n-benzyloxy carbonyl-l-aspartate,n-benzyloxycarbonyl-l-aspartic acid 1-methyl 4-tert-butyl ester,s-2-benzyloxycarbonylamino-succinic acid 4-tert-butyl ester 1-methyl ester
10593022
4-O-tert-butyl 1-O-methyl (2S)-2-(phenylmethoxycarbonylamino)butanedioate
CC(C)(C)OC(=O)CC(C(=O)OC)NC(=O)OCC1=CC=CC=C1
Beilstein 2601725
Formula Weight 337.37
Physical Form Powder
Percent Purity 98%
Color White
Quantity 25g
Chemical Name or Material N-Benzyloxycarbonyl-L-aspartic acid 4-tert-butyl ester 1-methyl ester

storageNote1 : -30° to -10°C