Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Benzyloxycarbonyl-L-glutamic acid 1-tert-butyl ester dicyclohexylammonium salt, 95%

Click to view available options
Quantity:
1g
5g
34897-61-5
C29H46N2O6
518.695
MFCD00077017
POQMGEMMYVJQOK-ZOWNYOTGSA-N
z-glu-otbu dcha,dicyclohexylamine s-4-benzyloxy carbonyl amino-5-tert-butoxy-5-oxopentanoate,z-glu otbu-oh?cha,z-glu-otbu.dcha,4s-4-benzyloxy carbonyl amino-5-tert-butoxy-5-oxopentanoic acid; dicha,n-alpha-benzyloxycarbonyl-l-glutamic acid alpha-t-butyl ester dicyclohexylamine,4s-4-benzyloxy carbonyl amino-5-tert-butoxy-5-oxopentanoic acid; n-cyclohexylcyclohexanamine
13474835
N-cyclohexylcyclohexanamine;(4S)-5-[(2-methylpropan-2-yl)oxy]-5-oxo-4-(phenylmethoxycarbonylamino)pentanoic acid
CC(C)(C)OC(=O)C(CCC(=O)O)NC(=O)OCC1=CC=CC=C1.C1CCC(CC1)NC2CCCCC2
34897-61-5
C29H46N2O6
518.695
MFCD00077017
POQMGEMMYVJQOK-ZOWNYOTGSA-N
z-glu-otbu dcha,dicyclohexylamine s-4-benzyloxy carbonyl amino-5-tert-butoxy-5-oxopentanoate,z-glu otbu-oh?cha,z-glu-otbu.dcha,4s-4-benzyloxy carbonyl amino-5-tert-butoxy-5-oxopentanoic acid; dicha,n-alpha-benzyloxycarbonyl-l-glutamic acid alpha-t-butyl ester dicyclohexylamine,4s-4-benzyloxy carbonyl amino-5-tert-butoxy-5-oxopentanoic acid; n-cyclohexylcyclohexanamine
13474835
N-cyclohexylcyclohexanamine;(4S)-5-[(2-methylpropan-2-yl)oxy]-5-oxo-4-(phenylmethoxycarbonylamino)pentanoic acid
CC(C)(C)OC(=O)C(CCC(=O)O)NC(=O)OCC1=CC=CC=C1.C1CCC(CC1)NC2CCCCC2
Beilstein 4286980
Formula Weight 518.69
Physical Form Solid
Percent Purity 95%
Quantity 1g
Chemical Name or Material N-Benzyloxycarbonyl-L-glutamic acid 1-tert-butyl ester dicyclohexylammonium salt

storageNote1 : -30° to -10°C