Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Benzyloxycarbonyl-L-glutamic acid 5-tert-butyl ester, 97%

Click to view available options
Quantity:
1g
5g
3886-08-6
C17H22NO6
336.37
MFCD00038274
GLMODRZPPBZPPB-ZDUSSCGKSA-M
z-glu otbu-oh,cbz-glu obut-oh,cbz-l-glu otbu-oh,s-2-benzyloxy carbonyl amino-5-tert-butoxy-5-oxopentanoic acid,n-cbz-l-glutamic acid 5-tert-butyl ester,n-benzyloxycarbonyl-l-glutamic acid 5-tert-butyl ester,2s-2-benzyloxy carbonyl amino-5-tert-butoxy-5-oxopentanoic acid,2s-4-tert-butyl oxycarbonyl-2-phenylmethoxy carbonylamino butanoic acid,5-tert-butyl n-carbobenzoxy-l-glutamate,pubchem6261
107472
(2S)-5-[(2-methylpropan-2-yl)oxy]-5-oxo-2-(phenylmethoxycarbonylamino)pentanoic acid
CC(C)(C)OC(=O)CCC(C(=O)O)NC(=O)OCC1=CC=CC=C1
3886-08-6
C17H22NO6
336.37
MFCD00038274
GLMODRZPPBZPPB-ZDUSSCGKSA-M
z-glu otbu-oh,cbz-glu obut-oh,cbz-l-glu otbu-oh,s-2-benzyloxy carbonyl amino-5-tert-butoxy-5-oxopentanoic acid,n-cbz-l-glutamic acid 5-tert-butyl ester,n-benzyloxycarbonyl-l-glutamic acid 5-tert-butyl ester,2s-2-benzyloxy carbonyl amino-5-tert-butoxy-5-oxopentanoic acid,2s-4-tert-butyl oxycarbonyl-2-phenylmethoxy carbonylamino butanoic acid,5-tert-butyl n-carbobenzoxy-l-glutamate,pubchem6261
107472
(2S)-5-[(2-methylpropan-2-yl)oxy]-5-oxo-2-(phenylmethoxycarbonylamino)pentanoic acid
CC(C)(C)OC(=O)CCC(C(=O)O)NC(=O)OCC1=CC=CC=C1
Optical Rotation −13° (c=2 in Methanol)
Quantity 1g
Melting Point 85°C to 88°C
Formula Weight 337.37
Percent Purity 97%
Chemical Name or Material N-Benzyloxycarbonyl-L-glutamic acid 5-tert-butyl ester

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning