Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Benzyloxycarbonyl-L-glutaminylglycine, 98%

Click to view available options
Quantity:
250mg
1g
5g
6610-42-0
C15H19N3O6
337.332
MFCD00055926
SOUXAAOTONMPRY-NSHDSACASA-N
z-gln-gly-oh,s-2-5-amino-2-benzyloxy carbonyl amino-5-oxopentanamido acetic acid,benzyloxycarbonyl-l-glutaminylglycine,cbz-l-gln-gly-oh,benzyloxy carbonyl-l-glutaminylglycine,s-2-5-amino-2-benzyloxy carbonyl-amino-5-oxopentanamido acetic acid,2s-2-benzyloxy carbonyl amino-4-carbamoylbutanamido acetic acid,2-2s-5-amino-5-oxo-2-phenylmethoxycarbonylamino pentanoyl amino acetic acid
6994993
2-[[(2S)-5-amino-5-oxo-2-(phenylmethoxycarbonylamino)pentanoyl]amino]acetic acid
C1=CC=C(C=C1)COC(=O)NC(CCC(=O)N)C(=O)NCC(=O)O
6610-42-0
C15H19N3O6
337.332
MFCD00055926
SOUXAAOTONMPRY-NSHDSACASA-N
z-gln-gly-oh,s-2-5-amino-2-benzyloxy carbonyl amino-5-oxopentanamido acetic acid,benzyloxycarbonyl-l-glutaminylglycine,cbz-l-gln-gly-oh,benzyloxy carbonyl-l-glutaminylglycine,s-2-5-amino-2-benzyloxy carbonyl-amino-5-oxopentanamido acetic acid,2s-2-benzyloxy carbonyl amino-4-carbamoylbutanamido acetic acid,2-2s-5-amino-5-oxo-2-phenylmethoxycarbonylamino pentanoyl amino acetic acid
6994993
2-[[(2S)-5-amino-5-oxo-2-(phenylmethoxycarbonylamino)pentanoyl]amino]acetic acid
C1=CC=C(C=C1)COC(=O)NC(CCC(=O)N)C(=O)NCC(=O)O
Beilstein 2179068
Formula Weight 337.33
Physical Form Solid
Percent Purity 98%
Quantity 5g
Chemical Name or Material N-Benzyloxycarbonyl-L-glutaminylglycine

storageNote1 : -30° to -10°C