Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Benzyloxycarbonyl-L-leucine N-succinimidyl ester, 95%

Click to view available options
Quantity:
5g
25g
100g
3397-35-1
C18H22N2O6
362.382
MFCD00042759
YHZUOMRURVTBMO-AWEZNQCLSA-N
z-leu-osu,n-carbobenzoxy-l-leucine n-succinimidyl ester,s-2,5-dioxopyrrolidin-1-yl 2-benzyloxy carbonyl amino-4-methylpentanoate,2,5-dioxopyrrolidin-1-yl 2s-2-benzyloxy carbonyl amino-4-methylpentanoate,2,5-dioxoazolidinyl 2s-4-methyl-2-phenylmethoxy carbonylamino pentanoate,z-l-leucine hydroxysuccinimide ester,n-cbz-l-leucine n-succinimidyl ester,z-l-leucine n-hydroxysuccinimide ester,n-cbz-l-leucine n-hydroxysuccinimide ester,z-l-leucine hydroxysuccinimide ester gr
11089751
(2,5-dioxopyrrolidin-1-yl) (2S)-4-methyl-2-(phenylmethoxycarbonylamino)pentanoate
CC(C)CC(C(=O)ON1C(=O)CCC1=O)NC(=O)OCC2=CC=CC=C2
3397-35-1
C18H22N2O6
362.382
MFCD00042759
YHZUOMRURVTBMO-AWEZNQCLSA-N
z-leu-osu,n-carbobenzoxy-l-leucine n-succinimidyl ester,s-2,5-dioxopyrrolidin-1-yl 2-benzyloxy carbonyl amino-4-methylpentanoate,2,5-dioxopyrrolidin-1-yl 2s-2-benzyloxy carbonyl amino-4-methylpentanoate,2,5-dioxoazolidinyl 2s-4-methyl-2-phenylmethoxy carbonylamino pentanoate,z-l-leucine hydroxysuccinimide ester,n-cbz-l-leucine n-succinimidyl ester,z-l-leucine n-hydroxysuccinimide ester,n-cbz-l-leucine n-hydroxysuccinimide ester,z-l-leucine hydroxysuccinimide ester gr
11089751
(2,5-dioxopyrrolidin-1-yl) (2S)-4-methyl-2-(phenylmethoxycarbonylamino)pentanoate
CC(C)CC(C(=O)ON1C(=O)CCC1=O)NC(=O)OCC2=CC=CC=C2
Beilstein 1555663
Formula Weight 362.38
Physical Form Solid
Percent Purity 95%
Color White
Melting Point 116°C to 120°C
Quantity 25g
Chemical Name or Material N-Benzyloxycarbonyl-L-leucine N-succinimidyl ester

storageNote1 : 2° to 8°C