Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Benzyloxycarbonyl-L-phenylalaninamide, 95%

Click to view available options
Quantity:
5g
25g
100g
4801-80-3
C17H18N2O3
298.342
MFCD00077028
HHBOFAIEPRHUSR-HNNXBMFYSA-N
s-benzyl 1-amino-1-oxo-3-phenylpropan-2-yl carbamate,z-phe-nh2,cbz-phe-nh2,ambotzzaa1235,z-phe-nh,z-l-phenylalanine amide,z-phe-nh2,n-alpha-carbobenzoxy-l-phenylalanine amide,n-alpha-benzyloxycarbonyl-l-phenylalanine amide,nalpha-phenylmethoxy carbonyl-l-phenylalaninamide
2755911
benzyl N-[(2S)-1-amino-1-oxo-3-phenylpropan-2-yl]carbamate
C1=CC=C(C=C1)CC(C(=O)N)NC(=O)OCC2=CC=CC=C2
4801-80-3
C17H18N2O3
298.342
MFCD00077028
HHBOFAIEPRHUSR-HNNXBMFYSA-N
s-benzyl 1-amino-1-oxo-3-phenylpropan-2-yl carbamate,z-phe-nh2,cbz-phe-nh2,ambotzzaa1235,z-phe-nh,z-l-phenylalanine amide,z-phe-nh2,n-alpha-carbobenzoxy-l-phenylalanine amide,n-alpha-benzyloxycarbonyl-l-phenylalanine amide,nalpha-phenylmethoxy carbonyl-l-phenylalaninamide
2755911
benzyl N-[(2S)-1-amino-1-oxo-3-phenylpropan-2-yl]carbamate
C1=CC=C(C=C1)CC(C(=O)N)NC(=O)OCC2=CC=CC=C2
Quantity 5g
Formula Weight 298.34
Physical Form Solid
Percent Purity 95%
Chemical Name or Material N-Benzyloxycarbonyl-L-phenylalaninamide

storageNote1 : 2° to 8°C