Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Benzyloxycarbonyl-L-phenylalanine N-succinimidyl ester, 95%

Click to view available options
Quantity:
5g
25g
100g
3397-32-8
C21H20N2O6
396.399
MFCD00053549
MOJNKVWQJKPXCT-KRWDZBQOSA-N
z-phe-osu,zpheosu,2,5-dioxopyrrolidin-1-yl 2s-2-benzyloxy carbonyl amino-3-phenylpropanoate,ambotzzaa1237,z-l-phe-osu,z-l-phenylalanine n-hydroxysuccinimide ester,n-benzyloxycarbonyl-l-phenylalanine succinimidyl ester,n-alpha-benzyloxycarbonyl-l-phenylalanine succinimidyl ester,n-benzyloxycarbonylphenylalanine-n-hydroxysuccinimide ester,2,5-dioxopyrrolidin-1-yl n-benzyloxy carbonyl-l-phenylalaninate
12876457
(2,5-dioxopyrrolidin-1-yl) (2S)-3-phenyl-2-(phenylmethoxycarbonylamino)propanoate
C1CC(=O)N(C1=O)OC(=O)C(CC2=CC=CC=C2)NC(=O)OCC3=CC=CC=C3
3397-32-8
C21H20N2O6
396.399
MFCD00053549
MOJNKVWQJKPXCT-KRWDZBQOSA-N
z-phe-osu,zpheosu,2,5-dioxopyrrolidin-1-yl 2s-2-benzyloxy carbonyl amino-3-phenylpropanoate,ambotzzaa1237,z-l-phe-osu,z-l-phenylalanine n-hydroxysuccinimide ester,n-benzyloxycarbonyl-l-phenylalanine succinimidyl ester,n-alpha-benzyloxycarbonyl-l-phenylalanine succinimidyl ester,n-benzyloxycarbonylphenylalanine-n-hydroxysuccinimide ester,2,5-dioxopyrrolidin-1-yl n-benzyloxy carbonyl-l-phenylalaninate
12876457
(2,5-dioxopyrrolidin-1-yl) (2S)-3-phenyl-2-(phenylmethoxycarbonylamino)propanoate
C1CC(=O)N(C1=O)OC(=O)C(CC2=CC=CC=C2)NC(=O)OCC3=CC=CC=C3
Beilstein 1557534
Formula Weight 396.4
Physical Form Solid
Percent Purity 95%
Quantity 25g
Chemical Name or Material N-Benzyloxycarbonyl-L-phenylalanine N-succinimidyl ester

storageNote1 : 2° to 8°C